missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromocoumarin-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1535414

 View more versions of this product

Catalog No. AAA1535414



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-Bromocoumarin-3-carboxylic acid
194°C to 196°C
MFCD00047640
13960
XFQHPAXNKDYMOX-UHFFFAOYSA-N
6-bromo-2-oxochromene-3-carboxylic acid
694463
97%
2199-87-3
C10H5BrO4
25 g
6-bromo-2-oxo-2h-chromene-3-carboxylic acid, 6-bromocoumarin-3-carboxylic acid, acmc-20alzr, maybridge1_000837, 6-bromo-3-carboxy-coumarin, 6-bromo-coumarin-3-carboxylic acid, 2-oxo-6-bromo-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylicacid,6-bromo-2-oxo, 2h-1-benzopyran-3-carboxylicacid, 6-bromo-2-oxo
C1=CC2=C(C=C1Br)C=C(C(=O)O2)C(=O)O
269.05
269.05

Chemical Identifiers

2199-87-3
269.05
XFQHPAXNKDYMOX-UHFFFAOYSA-N
694463
C1=CC2=C(C=C1Br)C=C(C(=O)O2)C(=O)O
C10H5BrO4
MFCD00047640
6-bromo-2-oxo-2h-chromene-3-carboxylic acid, 6-bromocoumarin-3-carboxylic acid, acmc-20alzr, maybridge1_000837, 6-bromo-3-carboxy-coumarin, 6-bromo-coumarin-3-carboxylic acid, 2-oxo-6-bromo-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylicacid,6-bromo-2-oxo, 2h-1-benzopyran-3-carboxylicacid, 6-bromo-2-oxo
6-bromo-2-oxochromene-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only