missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromocoumarin-3-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ A1535422

 View more versions of this product

Catalog No. AAA1535422



Specifications

Specifications

6-Bromocoumarin-3-carboxylic acid
100g
C10H5BrO4
13960
XFQHPAXNKDYMOX-UHFFFAOYSA-N
6-bromo-2-oxochromene-3-carboxylic acid
694463
97%
194°C to 196°C
2199-87-3
MFCD00047640
6-bromo-2-oxo-2h-chromene-3-carboxylic acid, 6-bromocoumarin-3-carboxylic acid, acmc-20alzr, maybridge1_000837, 6-bromo-3-carboxy-coumarin, 6-bromo-coumarin-3-carboxylic acid, 2-oxo-6-bromo-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylicacid,6-bromo-2-oxo, 2h-1-benzopyran-3-carboxylicacid, 6-bromo-2-oxo
C1=CC2=C(C=C1Br)C=C(C(=O)O2)C(=O)O
269.05
269.05

Chemical Identifiers

2199-87-3
269.05
XFQHPAXNKDYMOX-UHFFFAOYSA-N
694463
C1=CC2=C(C=C1Br)C=C(C(=O)O2)C(=O)O
C10H5BrO4
MFCD00047640
6-bromo-2-oxo-2h-chromene-3-carboxylic acid, 6-bromocoumarin-3-carboxylic acid, acmc-20alzr, maybridge1_000837, 6-bromo-3-carboxy-coumarin, 6-bromo-coumarin-3-carboxylic acid, 2-oxo-6-bromo-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylicacid,6-bromo-2-oxo, 2h-1-benzopyran-3-carboxylicacid, 6-bromo-2-oxo
6-bromo-2-oxochromene-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents