missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromochromone-3-carbonitrile, TCI America™

$111.53 - $330.00

Chemical Identifiers

CAS 52817-13-7
Molecular Formula C10H4BrNO2
Molecular Weight (g/mol) 250.051
MDL Number MFCD00191842
InChI Key MGVVCEKLWMZFLS-UHFFFAOYSA-N
Synonym 6-Bromo-3-cyanochromone, 6-Bromo-4-oxo-4H-1-benzopyran-3-carbonitrile
PubChem CID 703036
IUPAC Name 6-bromo-4-oxochromene-3-carbonitrile
SMILES C1=CC2=C(C=C1Br)C(=O)C(=CO2)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B49681G View Documents TCI America
B49681G
1g Each for $111.53
 
B49685G View Documents TCI America
B49685G
5g Each for $330.00
 
Specifications

Chemical Identifiers

52817-13-7
250.051
MGVVCEKLWMZFLS-UHFFFAOYSA-N
703036
C1=CC2=C(C=C1Br)C(=O)C(=CO2)C#N
C10H4BrNO2
MFCD00191842
6-Bromo-3-cyanochromone, 6-Bromo-4-oxo-4H-1-benzopyran-3-carbonitrile
6-bromo-4-oxochromene-3-carbonitrile

Specifications

Yellow
1g
C10H4BrNO2
3439
MGVVCEKLWMZFLS-UHFFFAOYSA-N
6-bromo-4-oxochromene-3-carbonitrile
703036
Crystal-Powder at 20°C
6-Bromochromone-3-carbonitrile
225°C
52817-13-7
MFCD00191842
6-Bromo-3-cyanochromone, 6-Bromo-4-oxo-4H-1-benzopyran-3-carbonitrile
C1=CC2=C(C=C1Br)C(=O)C(=CO2)C#N
250.051
250.05
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No