missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromochromone-2-carboxylic acid, 97%

$106.23 - $331.08

Chemical Identifiers

CAS 51484-06-1
Molecular Formula C10H5BrO4
Molecular Weight (g/mol) 269.05
MDL Number MFCD01242619
InChI Key QSBZDBNPXSVVHH-UHFFFAOYSA-N
Synonym 6-bromo-4-oxo-4h-1-benzopyran-2-carboxylic acid, 6-bromo-4-oxo-4h-chromene-2-carboxylic acid, 6-bromo-4-oxo-4h-chromene-2-carboxylicacid, 6-bromochromone-2-carboxylic acid, 6-bromochromone-2-carboxylicacid, 6-brornochrornone-2-carboxylic acid, acmc-20alzt
PubChem CID 688867
IUPAC Name 6-bromo-4-oxochromene-2-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C(=O)C=C(O2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3200906
View Documents
Alfa Aesar™
H3200906
5g Each for $106.23
 
AAH3200914
View Documents
Alfa Aesar™
H3200914
25g Each for $331.08
 
Specifications

Chemical Identifiers

51484-06-1
269.05
QSBZDBNPXSVVHH-UHFFFAOYSA-N
688867
C1=CC2=C(C=C1Br)C(=O)C=C(O2)C(=O)O
C10H5BrO4
MFCD01242619
6-bromo-4-oxo-4h-1-benzopyran-2-carboxylic acid, 6-bromo-4-oxo-4h-chromene-2-carboxylic acid, 6-bromo-4-oxo-4h-chromene-2-carboxylicacid, 6-bromochromone-2-carboxylic acid, 6-bromochromone-2-carboxylicacid, 6-brornochrornone-2-carboxylic acid, acmc-20alzt
6-bromo-4-oxochromene-2-carboxylic acid

Specifications

265°C
51484-06-1
MFCD01242619
QSBZDBNPXSVVHH-UHFFFAOYSA-N
6-bromo-4-oxochromene-2-carboxylic acid
688867
97%
5g
C10H5BrO4
6-bromo-4-oxo-4h-1-benzopyran-2-carboxylic acid, 6-bromo-4-oxo-4h-chromene-2-carboxylic acid, 6-bromo-4-oxo-4h-chromene-2-carboxylicacid, 6-bromochromone-2-carboxylic acid, 6-bromochromone-2-carboxylicacid, 6-brornochrornone-2-carboxylic acid, acmc-20alzt
C1=CC2=C(C=C1Br)C(=O)C=C(O2)C(=O)O
269.05
269.05
6-Bromochromone-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures