Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromoacetyl-1,4-benzodioxane

Chemical Identifiers

Chemical Name or Material 6-Bromoacetyl-1,4-benzodioxane
CAS 4629-54-3
Molecular Formula C10H9BrO3
Formula Weight 257.08
MDL Number MFCD00099402
UN Number UN1759
Synonym 3',4'-Ethylenedioxy-2-bromoacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5069606

 
alfa aesar™
H5069606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5069603

 
alfa aesar™
H5069603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 257.08
Chemical Name or Material 6-Bromoacetyl-1,4-benzodioxane
Molecular Formula C10H9BrO3
MDL Number MFCD00099402
CAS 4629-54-3
UN Number UN1759
Synonym 3',4'-Ethylenedioxy-2-bromoacetophenone
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

Applications
6-Bromoacetyl-1,4-benzodioxane is used as an intermediate in pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.