missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-8-iodoimidazo[1,2-a]pyridine, 95%

Manufacturer:  Alfa Aesar™ H3754206

 View more versions of this product

Catalog No. AAH3754206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

6-Bromo-8-iodoimidazo[1,2-a]pyridine
Yellow
5g
C7H4BrIN2
6-bromo-8-iodoimidazo 1,2-a pyridine, 6-bromo-8-iodo-imidazo 1,2-a pyridine, 6-bromo-8-iodoimidazo 1,2-a pyridine,, imidazo 1,2-a pyridine,6-bromo-8-iodo, imidazo 1,2-a pyridine, 6-bromo-8-iodo
C1=CN2C=C(C=C(C2=N1)I)Br
322.931
322.92
95%
Light Sensitive
147°C to 150°C
1364917-14-5
MFCD21609484
OKEMQQRZPHOOGS-UHFFFAOYSA-N
6-bromo-8-iodoimidazo[1,2-a]pyridine
71299170
Powder

Chemical Identifiers

1364917-14-5
322.931
OKEMQQRZPHOOGS-UHFFFAOYSA-N
71299170
C1=CN2C=C(C=C(C2=N1)I)Br
C7H4BrIN2
MFCD21609484
6-bromo-8-iodoimidazo 1,2-a pyridine, 6-bromo-8-iodo-imidazo 1,2-a pyridine, 6-bromo-8-iodoimidazo 1,2-a pyridine,, imidazo 1,2-a pyridine,6-bromo-8-iodo, imidazo 1,2-a pyridine, 6-bromo-8-iodo
6-bromo-8-iodoimidazo[1,2-a]pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents