missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromo-8-fluoroquinoline, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC452560010

 View more versions of this product

Catalog No. AC452560010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

6-Bromo-8-fluoroquinoline
103°C to 106°C
220513-46-2
C9H5BrFN
DDCOYWOYIUEOHQ-UHFFFAOYSA-N
6-bromo-8-fluoroquinoline
46738885
97%
Glass Bottle
1g
96% min. (GC)
6-bromo-8-fluoro-1-azanaphthalene, 6-bromo-8-fluoro-quinoline, 6-bromo-8-fluoroquinoline, acmc-1crnu
C1=CC2=CC(=CC(=C2N=C1)F)Br
226.05
226.05

Chemical Identifiers

220513-46-2
226.05
6-bromo-8-fluoro-1-azanaphthalene, 6-bromo-8-fluoro-quinoline, 6-bromo-8-fluoroquinoline, acmc-1crnu
6-bromo-8-fluoroquinoline
C9H5BrFN
DDCOYWOYIUEOHQ-UHFFFAOYSA-N
46738885
C1=CC2=CC(=CC(=C2N=C1)F)Br
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents