Please login to your online account to display your discounted pricing

6-Bromo-8-fluoroquinoline, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 6-Bromo-8-fluoroquinoline
Assay Percent Range 97%
Molecular Formula C9H5BrFN
Formula Weight 226.05
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC452560010

  SDS
thermo fisher scientific
AC452560010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC452560050

  SDS
thermo fisher scientific
AC452560050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 6-Bromo-8-fluoroquinoline
CAS 220513-46-2
Assay Percent Range 97%
Quantity 1g
Molecular Formula C9H5BrFN
Formula Weight 226.05