Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine, 96%

Chemical Identifiers

Chemical Name or Material 6-Bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine
CAS 1160102-28-2
Formula Weight 232.05
MDL Number MFCD20441785
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6225903

  SDS
alfa aesar™
H6225903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62259MD

  SDS
alfa aesar™
H62259MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 232.05
Quantity 1g
Physical Form Solid
CAS 1160102-28-2
MDL Number MFCD20441785
Chemical Name or Material 6-Bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine
Assay 96%