missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one, 96%

Manufacturer:  Alfa Aesar™ H6202603

 View more versions of this product

Catalog No. AAH6202603



Specifications

Specifications

6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one
355423-58-4
MFCD20441932
AVHJMSMGASUDIT-UHFFFAOYSA-N
6-bromo-7-fluoro-4H-1,4-benzoxazin-3-one
23574320
Solid
1g
C8H5BrFNO2
2h-1,4-benzoxazin-3 4h-one, 6-bromo-7-fluoro
C1C(=O)NC2=CC(=C(C=C2O1)F)Br
246.035
246.03
96%

Chemical Identifiers

355423-58-4
246.035
AVHJMSMGASUDIT-UHFFFAOYSA-N
23574320
C1C(=O)NC2=CC(=C(C=C2O1)F)Br
C8H5BrFNO2
MFCD20441932
2h-1,4-benzoxazin-3 4h-one, 6-bromo-7-fluoro
6-bromo-7-fluoro-4H-1,4-benzoxazin-3-one
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents