Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-7-azaindole, 96%

Chemical Identifiers

Chemical Name or Material 6-Bromo-7-azaindole
CAS 143468-13-7
Formula Weight 197.04
MDL Number MFCD06659667
UN Number UN2811
Beilstein 5424619
Synonym 6-Bromo-1H-pyrrolo[2,3-b]pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6686303

 
alfa aesar™
H6686303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6686306

 
alfa aesar™
H6686306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 197.04
Melting Point 192° to 195°C
Chemical Name or Material 6-Bromo-7-azaindole
Physical Form Solid
Assay 96%
CAS 143468-13-7
Health Hazard 2 GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
MDL Number MFCD06659667
UN Number UN2811
Beilstein 5424619
Color Off-white to yellow
Quantity 1g
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Synonym 6-Bromo-1H-pyrrolo[2,3-b]pyridine
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.