Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-5-methylpyridine-3-boronic acid, 95%

$241.08 - $554.49

Chemical Identifiers

Chemical Name or Material 6-Bromo-5-methylpyridine-3-boronic acid
CAS 1003043-34-2
Molecular Formula C6H7BBrNO2
Formula Weight 215.84
MDL Number MFCD08274468
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2722103

  SDS
alfa aesar™
H2722103
1g Each for $554.49

AAH27221MD

  SDS
alfa aesar™
H27221MD
250mg Each for $241.08
Description & Specifications

Specifications

Formula Weight 215.84
MDL Number MFCD08274468
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 1003043-34-2
Assay 95%
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Molecular Formula C6H7BBrNO2
Chemical Name or Material 6-Bromo-5-methylpyridine-3-boronic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.