missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-4-methyl-3-nitroquinoline, 96%

Manufacturer:  Alfa Aesar™ H54769MD

 View more versions of this product

Catalog No. AAH54769MD



Description

Description

Applications
6-Bromo-4-methyl-3-nitroquinoline is used as pharmaceutical intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
Specifications

Specifications

6-Bromo-4-methyl-3-nitroquinoline
250mg
C10H7BrN2O2
19927450
CC1=C2C=C(C=CC2=NC=C1[N+](=O)[O-])Br
267.082
267.08
152°C to 155°C
1185310-49-9
MFCD12761249
MYJZQOHLSJFXDS-UHFFFAOYSA-N
6-bromo-4-methyl-3-nitroquinoline
45789224
96%

Chemical Identifiers

1185310-49-9
267.082
MYJZQOHLSJFXDS-UHFFFAOYSA-N
6-bromo-4-methyl-3-nitroquinoline
C10H7BrN2O2
MFCD12761249
45789224
CC1=C2C=C(C=CC2=NC=C1[N+](=O)[O-])Br
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2° to 8°C

Documents