missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-4-chloroquinoline, 96%

$82.92 - $305.36

Chemical Identifiers

CAS 65340-70-7
Molecular Formula C9H5BrClN
Molecular Weight (g/mol) 242.5
MDL Number MFCD00511001
InChI Key KJILYZMXTLCPDQ-UHFFFAOYSA-N
Synonym 4-chloro-6-bromoquinoline, 4-choro-6-bromoquinoline, 6-bromo-4-chloro quinoline, 6-bromo-4-chloro-quinoline, 6-bromo-4-chloroquinoline, acmc-1bdfp, ksc493o7p, pubchem20825, quinoline, 6-bromo-4-chloro
PubChem CID 5139537
IUPAC Name 6-bromo-4-chloroquinoline
SMILES C1=CC2=NC=CC(=C2C=C1Br)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54902MD
View Documents
Alfa Aesar™
H54902MD
250mg Each for $82.92
 
AAH5490203
View Documents
Alfa Aesar™
H5490203
1g Each for $305.36
 
Specifications

Chemical Identifiers

65340-70-7
242.5
KJILYZMXTLCPDQ-UHFFFAOYSA-N
5139537
C1=CC2=NC=CC(=C2C=C1Br)Cl
C9H5BrClN
MFCD00511001
4-chloro-6-bromoquinoline, 4-choro-6-bromoquinoline, 6-bromo-4-chloro quinoline, 6-bromo-4-chloro-quinoline, 6-bromo-4-chloroquinoline, acmc-1bdfp, ksc493o7p, pubchem20825, quinoline, 6-bromo-4-chloro
6-bromo-4-chloroquinoline

Specifications

110°C to 112°C
65340-70-7
MFCD00511001
KJILYZMXTLCPDQ-UHFFFAOYSA-N
6-bromo-4-chloroquinoline
5139537
96%
250mg
C9H5BrClN
4-chloro-6-bromoquinoline, 4-choro-6-bromoquinoline, 6-bromo-4-chloro quinoline, 6-bromo-4-chloro-quinoline, 6-bromo-4-chloroquinoline, acmc-1bdfp, ksc493o7p, pubchem20825, quinoline, 6-bromo-4-chloro
C1=CC2=NC=CC(=C2C=C1Br)Cl
242.5
242.5
6-Bromo-4-chloroquinoline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen