missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-4-chloro-3-nitroquinoline, 96%

$121.19 - $356.80

Chemical Identifiers

CAS 723281-72-9
Molecular Formula C9H4BrClN2O2
Molecular Weight (g/mol) 287.497
MDL Number MFCD09909913
InChI Key KHKPVEMMXJCWGP-UHFFFAOYSA-N
Synonym 6-bromo-4-chloro-3-nitro-quinoline
PubChem CID 21984128
IUPAC Name 6-bromo-4-chloro-3-nitroquinoline
SMILES C1=CC2=NC=C(C(=C2C=C1Br)Cl)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54180MD View Documents Alfa Aesar™
H54180MD
250mg Each for $121.19
 
AAH5418003 View Documents Alfa Aesar™
H5418003
1g Each for $356.80
 
Specifications

Chemical Identifiers

723281-72-9
287.497
KHKPVEMMXJCWGP-UHFFFAOYSA-N
21984128
C1=CC2=NC=C(C(=C2C=C1Br)Cl)[N+](=O)[O-]
C9H4BrClN2O2
MFCD09909913
6-bromo-4-chloro-3-nitro-quinoline
6-bromo-4-chloro-3-nitroquinoline

Specifications

250mg
C9H4BrClN2O2
6-bromo-4-chloro-3-nitro-quinoline
C1=CC2=NC=C(C(=C2C=C1Br)Cl)[N+](=O)[O-]
287.497
287.5
6-Bromo-4-chloro-3-nitroquinoline
723281-72-9
MFCD09909913
KHKPVEMMXJCWGP-UHFFFAOYSA-N
6-bromo-4-chloro-3-nitroquinoline
21984128
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures