missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-4-chloro-3-nitroquinoline, 96%

Manufacturer:  Alfa Aesar™ H54180MD

 View more versions of this product

Catalog No. AAH54180MD



Specifications

Specifications

6-Bromo-4-chloro-3-nitroquinoline
723281-72-9
MFCD09909913
KHKPVEMMXJCWGP-UHFFFAOYSA-N
6-bromo-4-chloro-3-nitroquinoline
21984128
96%
250mg
C9H4BrClN2O2
6-bromo-4-chloro-3-nitro-quinoline
C1=CC2=NC=C(C(=C2C=C1Br)Cl)[N+](=O)[O-]
287.497
287.5

Chemical Identifiers

723281-72-9
287.497
KHKPVEMMXJCWGP-UHFFFAOYSA-N
21984128
C1=CC2=NC=C(C(=C2C=C1Br)Cl)[N+](=O)[O-]
C9H4BrClN2O2
MFCD09909913
6-bromo-4-chloro-3-nitro-quinoline
6-bromo-4-chloro-3-nitroquinoline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents