Learn More
6-Bromo-3-pyridinecarboxaldehyde, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 399850050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
6-Bromo-3-pyridinecarboxaldehyde | |
94% min. (GC) | |
C6H4BrNO | |
5g | |
PVUKGNBRJFTFNJ-UHFFFAOYSA-N | |
6-bromopyridine-3-carbaldehyde | |
11769234 | |
95% |
149806-06-4 | |
Glass bottle | |
MFCD04115419 | |
6-bromonicotinaldehyde, 2-bromo-5-formylpyridine, 2-bromopyridine-5-carbaldehyde, 6-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-bromo, 6-bromo-pyridine-3-carbaldehyde, 6-bromopyridine-3-carboxaldehyde, 2-bromo-5-pyridinecarbaldehyde, 6-bromo-3-formylpyridine | |
BrC1=CC=C(C=O)C=N1 | |
186.01 | |
186 |
Chemical Identifiers
149806-06-4 | |
186.01 | |
PVUKGNBRJFTFNJ-UHFFFAOYSA-N | |
11769234 | |
BrC1=CC=C(C=O)C=N1 |
C6H4BrNO | |
MFCD04115419 | |
6-bromonicotinaldehyde, 2-bromo-5-formylpyridine, 2-bromopyridine-5-carbaldehyde, 6-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-bromo, 6-bromo-pyridine-3-carbaldehyde, 6-bromopyridine-3-carboxaldehyde, 2-bromo-5-pyridinecarbaldehyde, 6-bromo-3-formylpyridine | |
6-bromopyridine-3-carbaldehyde |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only