Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-3-iodo-2,4-dimethylpyridine, 95%

$130.47 - $293.82

Chemical Identifiers

Chemical Name or Material 6-Bromo-3-iodo-2,4-dimethylpyridine
CAS 942206-05-5
Molecular Formula C7H7BrIN
Formula Weight 311.95
MDL Number MFCD09037470
Synonym 6-Bromo-2,4-dimethyl-3-iodopyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2703303

  SDS
alfa aesar™
H2703303
1g Each for $293.82

AAH27033MD

  SDS
alfa aesar™
H27033MD
250mg Each for $130.47
Description & Specifications

Specifications

Formula Weight 311.95
Molecular Formula C7H7BrIN
Melting Point 89° to 91°C
Synonym 6-Bromo-2,4-dimethyl-3-iodopyridine
MDL Number MFCD09037470
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
CAS 942206-05-5
Health Hazard 1 Danger
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 6-Bromo-3-iodo-2,4-dimethylpyridine
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.