Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-3-butyrylcoumarin, 97%

$74.58 - $263.69

Chemical Identifiers

Chemical Name or Material 6-Bromo-3-butyrylcoumarin
CAS 2199-83-9
Molecular Formula C13H11BrO2
Formula Weight 279.14
MDL Number MFCD00052894
Synonym 6-Bromo-3-butyryl-2H-chromen-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2500303

  SDS
alfa aesar™
B2500303
1g Each for $74.58

AAB2500306

  SDS
alfa aesar™
B2500306
5g Each for $263.69
Description & Specifications

Specifications

Formula Weight 279.14
Melting Point 163° to 165°C
Chemical Name or Material 6-Bromo-3-butyrylcoumarin
Molecular Formula C13H11BrO2
Assay 97%
CAS 2199-83-9
MDL Number MFCD00052894
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Synonym 6-Bromo-3-butyryl-2H-chromen-2-one
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.