missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-3,4-dihydro-2H-1,4-benzoxazine, 97%

$106.26 - $337.73

Chemical Identifiers

CAS 105655-01-4
Molecular Formula C8H8BrNO
Molecular Weight (g/mol) 214.062
MDL Number MFCD08544341
InChI Key RWKBNMSHIJBNAO-UHFFFAOYSA-N
Synonym 6-bromo-3,4-dihydro-2h-benzo b 1,4 oxazine, 6-bromo-3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 6-bromo-3,4-dihydro, 2h-1,4-benzoxazine,6-bromo-3,4-dihydro, pubchem17444, acmc-2098ha, 6-bromo-3,4-dihydro-2h-benzo 1,4 oxazinehydrochloride, 6-bromo-3,4-dihydro-2h-benzo 1,4oxazine, 6-bromo-3,4-dihydro-2h-benzo 1,4 oxazine hydrochlo
PubChem CID 10727336
IUPAC Name 6-bromo-3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=C(N1)C=C(C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61070MD View Documents Alfa Aesar™
H61070MD
250mg Each for $106.26
 
AAH6107003 View Documents Alfa Aesar™
H6107003
1g Each for $337.73
 
Specifications

Chemical Identifiers

105655-01-4
214.062
RWKBNMSHIJBNAO-UHFFFAOYSA-N
10727336
C1COC2=C(N1)C=C(C=C2)Br
C8H8BrNO
MFCD08544341
6-bromo-3,4-dihydro-2h-benzo b 1,4 oxazine, 6-bromo-3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 6-bromo-3,4-dihydro, 2h-1,4-benzoxazine,6-bromo-3,4-dihydro, pubchem17444, acmc-2098ha, 6-bromo-3,4-dihydro-2h-benzo 1,4 oxazinehydrochloride, 6-bromo-3,4-dihydro-2h-benzo 1,4oxazine, 6-bromo-3,4-dihydro-2h-benzo 1,4 oxazine hydrochlo
6-bromo-3,4-dihydro-2H-1,4-benzoxazine

Specifications

250mg
C8H8BrNO
6-bromo-3,4-dihydro-2h-benzo b 1,4 oxazine, 6-bromo-3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 6-bromo-3,4-dihydro, 2h-1,4-benzoxazine,6-bromo-3,4-dihydro, pubchem17444, acmc-2098ha, 6-bromo-3,4-dihydro-2h-benzo 1,4 oxazinehydrochloride, 6-bromo-3,4-dihydro-2h-benzo 1,4oxazine, 6-bromo-3,4-dihydro-2h-benzo 1,4 oxazine hydrochlo
C1COC2=C(N1)C=C(C=C2)Br
214.062
214.06
6-Bromo-3,4-dihydro-2H-1,4-benzoxazine
105655-01-4
MFCD08544341
RWKBNMSHIJBNAO-UHFFFAOYSA-N
6-bromo-3,4-dihydro-2H-1,4-benzoxazine
10727336
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning