missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-3-(3-oxycyclopentyl)indole, 95%

$197.39 - $471.89

Chemical Identifiers

CAS 676170-06-2
Molecular Formula C13H12BrNO
Molecular Weight (g/mol) 278.149
InChI Key TWDVYIPKPMKBCL-UHFFFAOYSA-N
Synonym 3-6-bromo-1h-indol-3-yl cyclopentanone, 3-6-bromo-1h-indol-3-yl cyclopentan-1-one
PubChem CID 22139468
IUPAC Name 3-(6-bromo-1H-indol-3-yl)cyclopentan-1-one
SMILES C1CC(=O)CC1C2=CNC3=C2C=CC(=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63858MD View Documents Alfa Aesar™
H63858MD
250mg Each for $197.39
 
AAH6385803 View Documents Alfa Aesar™
H6385803
1g Each for $471.89
 
Specifications

Chemical Identifiers

676170-06-2
278.149
3-6-bromo-1h-indol-3-yl cyclopentanone, 3-6-bromo-1h-indol-3-yl cyclopentan-1-one
3-(6-bromo-1H-indol-3-yl)cyclopentan-1-one
C13H12BrNO
TWDVYIPKPMKBCL-UHFFFAOYSA-N
22139468
C1CC(=O)CC1C2=CNC3=C2C=CC(=C3)Br

Specifications

250mg
C13H12BrNO
3-6-bromo-1h-indol-3-yl cyclopentanone, 3-6-bromo-1h-indol-3-yl cyclopentan-1-one
C1CC(=O)CC1C2=CNC3=C2C=CC(=C3)Br
278.149
278.14
95%
676170-06-2
21019370
TWDVYIPKPMKBCL-UHFFFAOYSA-N
3-(6-bromo-1H-indol-3-yl)cyclopentan-1-one
22139468
Solid
6-Bromo-3-(3-oxycyclopentyl)indole