missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromo-2-naphthol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC107171000

 View more versions of this product

Catalog No. AC107171000


 

Specifications Description & Specifications

Specifications

6-Bromo-2-naphthol
96.0
Authentic
125° to 130°C
Powder
≥96.0%
C10H7BrO
223.07
06, 651
15231-91-1
100.0
Glass bottle
Off-white to beige
100g
97%
BrC10H6OH
MFCD00004081

Chemical Identifiers

15231-91-1
100.0
C10H7BrO
223.07
6-Bromo-2-naphthol
96.0
97%
BrC10H6OH
MFCD00004081
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.