Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-2-methylquinoline, 97%

$94.35 - $307.85

Chemical Identifiers

Chemical Name or Material 6-Bromo-2-methylquinoline
CAS 877-42-9
Molecular Formula C10H8BrN
Formula Weight 222.08
MDL Number MFCD00079724
Synonym 6-Bromoquinaldine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2744706

 
alfa aesar™
H2744706
5g Each for $94.35

AAH2744714

 
alfa aesar™
H2744714
25g Each for $307.85
Description & Specifications

Specifications

Formula Weight 222.08
Chemical Name or Material 6-Bromo-2-methylquinoline
Assay 97%
Molecular Formula C10H8BrN
MDL Number MFCD00079724
CAS 877-42-9
Melting Point 100° to 103°C
Synonym 6-Bromoquinaldine
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.