Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-2-methylpyridine-3-boronic acid, 95%

$160.07 - $511.11

Chemical Identifiers

Chemical Name or Material 6-Bromo-2-methylpyridine-3-boronic acid
CAS 1072944-22-9
Molecular Formula C6H7BBrNO2
Formula Weight 215.84g/mol
MDL Number MFCD09037490
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2771503

 
alfa aesar™
H27715-03
1g Each for $511.11

AAH27715MD

 
alfa aesar™
H27715-MD
250mg Each for $160.07
Description & Specifications

Specifications

Melting Point ≈170°C
Quantity 1g
Chemical Name or Material 6-Bromo-2-methylpyridine-3-boronic acid
CAS 1072944-22-9
Assay 95%
Molecular Formula C6H7BBrNO2
Formula Weight 215.84g/mol
MDL Number MFCD09037490
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.