missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-2-methyl-4H-3,1-benzoxazin-4-one

$102.05 - $102.05

Chemical Identifiers

CAS 19165-25-4
Molecular Formula C9H6BrNO2
Formula Weight 240.06
MDL Number MFCD00115891
Chemical Name or Material 6-Bromo-2-methyl-4H-3,1-benzoxazin-4-one
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH50585MD SDS Alfa Aesar™
H50585MD
250mg Each for $102.05
AAH5058503 SDS Alfa Aesar™
H5058503
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

19165-25-4
240.06
6-Bromo-2-methyl-4H-3,1-benzoxazin-4-one
C9H6BrNO2
MFCD00115891

Specifications

19165-25-4
240.06
250mg
C9H6BrNO2
MFCD00115891
6-Bromo-2-methyl-4H-3,1-benzoxazin-4-one