missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromo-2-methyl-2H-indazole, 97%, ACROS Organics™

Chemical Identifiers

CAS 590417-95-1
Molecular Formula C8H7BrN2
Molecular Weight (g/mol) 211.06
InChI Key BVYFYDANLZQCPV-UHFFFAOYSA-N
Synonym 2-methyl-6-bromoindazole, 2h-indazole, 6-bromo-2-methyl, 2h-indazole,6-bromo-2-methyl, 6-bromo-2-methyl-2h-indazole, 6-bromo-2-methyl-indazole, acmc-209m9q, boron-mol bm788, pubchem20671
PubChem CID 22558624
IUPAC Name 6-bromo-2-methylindazole
SMILES CN1C=C2C=CC(=CC2=N1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444610010
SDS View Documents
Thermo Fisher Scientific
AC444610010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC444610050
SDS View Documents
Thermo Fisher Scientific
AC444610050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

590417-95-1
211.06
2-methyl-6-bromoindazole, 2h-indazole, 6-bromo-2-methyl, 2h-indazole,6-bromo-2-methyl, 6-bromo-2-methyl-2h-indazole, 6-bromo-2-methyl-indazole, acmc-209m9q, boron-mol bm788, pubchem20671
6-bromo-2-methylindazole
C8H7BrN2
BVYFYDANLZQCPV-UHFFFAOYSA-N
22558624
CN1C=C2C=CC(=CC2=N1)Br

Specifications

Glass Bottle
1g
96% min. (GC)
2-methyl-6-bromoindazole, 2h-indazole, 6-bromo-2-methyl, 2h-indazole,6-bromo-2-methyl, 6-bromo-2-methyl-2h-indazole, 6-bromo-2-methyl-indazole, acmc-209m9q, boron-mol bm788, pubchem20671
CN1C=C2C=CC(=CC2=N1)Br
211.06
211.06
6-Bromo-2-methyl-2H-indazole
85°C to 87°C
590417-95-1
C8H7BrN2
BVYFYDANLZQCPV-UHFFFAOYSA-N
6-bromo-2-methylindazole
22558624
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning