missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Bromo-2-methyl-2H-indazole, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444610050
Specifications
6-Bromo-2-methyl-2H-indazole | |
5g | |
96% min. (GC) | |
2-methyl-6-bromoindazole, 2h-indazole, 6-bromo-2-methyl, 2h-indazole,6-bromo-2-methyl, 6-bromo-2-methyl-2h-indazole, 6-bromo-2-methyl-indazole, acmc-209m9q, boron-mol bm788, pubchem20671 | |
CN1C=C2C=CC(=CC2=N1)Br | |
211.06 | |
211.06 |
85°C to 87°C | |
590417-95-1 | |
C8H7BrN2 | |
BVYFYDANLZQCPV-UHFFFAOYSA-N | |
6-bromo-2-methylindazole | |
22558624 | |
97% |
Chemical Identifiers
590417-95-1 | |
211.06 | |
2-methyl-6-bromoindazole, 2h-indazole, 6-bromo-2-methyl, 2h-indazole,6-bromo-2-methyl, 6-bromo-2-methyl-2h-indazole, 6-bromo-2-methyl-indazole, acmc-209m9q, boron-mol bm788, pubchem20671 | |
6-bromo-2-methylindazole |
C8H7BrN2 | |
BVYFYDANLZQCPV-UHFFFAOYSA-N | |
22558624 | |
CN1C=C2C=CC(=CC2=N1)Br |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning