missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-2-methoxyquinoline, 96%

Manufacturer:  Alfa Aesar™ H5422606

 View more versions of this product

Catalog No. AAH5422606



Specifications

Specifications

6-Bromo-2-methoxyquinoline
5g
C10H8BrNO
2-methoxy-6-bromoquinoline
COC1=NC2=C(C=C1)C=C(C=C2)Br
238.084
238.08
87°C to 89°C
99455-05-7
MFCD11847805
KBTKKEMYFUMFSJ-UHFFFAOYSA-N
6-bromo-2-methoxyquinoline
10657538
96%

Chemical Identifiers

99455-05-7
238.084
KBTKKEMYFUMFSJ-UHFFFAOYSA-N
10657538
COC1=NC2=C(C=C1)C=C(C=C2)Br
C10H8BrNO
MFCD11847805
2-methoxy-6-bromoquinoline
6-bromo-2-methoxyquinoline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents