missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-2-methoxyquinoline, 96%

Manufacturer:  Alfa Aesar™ H5422603

 View more versions of this product

Catalog No. AAH5422603



Description

Description

Specifications

Specifications

6-Bromo-2-methoxyquinoline
1g
C10H8BrNO
2-methoxy-6-bromoquinoline, 6-bromanyl-2-methoxy-quinoline, 6-bromo-2-methoxy-quinoline, quinoline, 6-bromo-2-methoxy
COC1=NC2=C(C=C1)C=C(C=C2)Br
238.084
238.08
87°C to 89°C
99455-05-7
MFCD11847805
KBTKKEMYFUMFSJ-UHFFFAOYSA-N
6-bromo-2-methoxyquinoline
10657538
96%

Chemical Identifiers

99455-05-7
238.084
KBTKKEMYFUMFSJ-UHFFFAOYSA-N
10657538
COC1=NC2=C(C=C1)C=C(C=C2)Br
C10H8BrNO
MFCD11847805
2-methoxy-6-bromoquinoline, 6-bromanyl-2-methoxy-quinoline, 6-bromo-2-methoxy-quinoline, quinoline, 6-bromo-2-methoxy
6-bromo-2-methoxyquinoline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents