missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Bromo-2-hydroxy-3-methoxybenzaldehyde 99.0+%, TCI America™
Supplier: TCI America B148725G
Specifications
6-Bromo-2-hydroxy-3-methoxybenzaldehyde | |
103°C | |
C8H7BrO3 | |
25 g | |
COC1=CC=C(Br)C(C=O)=C1O | |
231.05 | |
231.05 | |
Crystalline Powder |
20035-41-0 | |
Yellow | |
MFCD00046152 | |
JUPJZUTYDWXZAQ-UHFFFAOYSA-N | |
6-bromo-2-hydroxy-3-methoxybenzaldehyde | |
262237 | |
≥99.0% (GC,T) |
Chemical Identifiers
20035-41-0 | |
231.05 | |
JUPJZUTYDWXZAQ-UHFFFAOYSA-N | |
6-bromo-2-hydroxy-3-methoxybenzaldehyde |
C8H7BrO3 | |
MFCD00046152 | |
262237 | |
COC1=CC=C(Br)C(C=O)=C1O |
Safety and Handling
TSCA : No