Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-2'-chloro-2,3'-bipyridine, 95%

$76.85 - $277.24

Chemical Identifiers

Chemical Name or Material 6-Bromo-2'-chloro-2,3'-bipyridine
CAS 942206-02-2
Molecular Formula C10H6BrClN2
Formula Weight 269.53
MDL Number MFCD09037439
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27149MC

  SDS
alfa aesar™
H27149MC
100mg Each for $76.85

AAH27149ME

  SDS
alfa aesar™
H27149ME
500mg Each for $277.24
Description & Specifications

Specifications

Formula Weight 269.53
Chemical Name or Material 6-Bromo-2'-chloro-2,3'-bipyridine
Assay 95%
Molecular Formula C10H6BrClN2
MDL Number MFCD09037439
CAS 942206-02-2
Melting Point 139° to 142°C
Quantity 100mg
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.