Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-2-chloro-1,8-naphthyridine, 96%

$88.59 - $304.36

Chemical Identifiers

Chemical Name or Material 6-Bromo-2-chloro-1,8-naphthyridine
CAS 902837-40-5
Molecular Formula C8H4BrClN2
Formula Weight 243.49
MDL Number MFCD07781152
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5004303

  SDS
alfa aesar™
H5004303
1g Each for $304.36

AAH50043MD

  SDS
alfa aesar™
H50043MD
250mg Each for $88.59
Description & Specifications

Specifications

Formula Weight 243.49
Quantity 1g
Molecular Formula C8H4BrClN2
Chemical Name or Material 6-Bromo-2-chloro-1,8-naphthyridine
MDL Number MFCD07781152
CAS 902837-40-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Assay 96%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
6-Bromo-2-chloro-1,8-naphthyridine, is used as raw material for synthesis of pharmaceutical intermediates.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.