Please login to your online account to display your discounted pricing

6-Bromo-2-benzothiazolinone, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  33988-0050

 View more versions of this product

Catalog No. AC339880050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 230.08
Physical Form Crystalline Powder
Quantity 5g
Color White to off-white
Melting Point 228° to 232°C
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 6-Bromo-2-benzothiazolinone
Name Note 98%
CAS Min % 97.5
Assay Percent Range 98%
MDL Number MFCD00239363
CAS 62266-82-4
Molecular Formula C7 H4 Br N O S
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 6-Bromo-2-benzothiazolinone
Name Note 98%
CAS 62266-82-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7 H4 Br N O S
Formula Weight 230.08
MDL Number MFCD00239363