Please login to your online account to display your discounted pricing

6-Bromo-2-benzothiazolinone, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  33988-0010

 View more versions of this product

Catalog No. AC339880010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 230.08
Quantity 1g
Color White to off-white
Melting Point 228° to 232°C
CAS Max % 100.0
Name Note 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C7 H4 Br N O S
CAS 62266-82-4
CAS Min % 97.5
Chemical Name or Material 6-Bromo-2-benzothiazolinone
MDL Number MFCD00239363
Assay Percent Range 98%

Specifications

Chemical Name or Material 6-Bromo-2-benzothiazolinone
Name Note 98%
CAS 62266-82-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7 H4 Br N O S
Formula Weight 230.08
MDL Number MFCD00239363