missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
6-Bromo-2-benzothiazolinone, 98%, ACROS Organics™
Chemical Identifiers
CAS |
62266-82-4 |
Molecular Formula |
C7H4BrNOS |
Molecular Weight (g/mol) |
230.08 |
MDL Number |
MFCD00239363 |
InChI Key |
HECJMTPEVWQFCY-UHFFFAOYSA-N |
Synonym |
2 3h-benzothiazolone, 6-bromo, 2-benzothiazolol, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 6-bromo-2-benzothiazolinone, 6-bromo-2-hydroxy benzothiazol, 6-bromo-3h-benzothiazol-2-one, 6-bromobenzo d thiazol-2 3h-one, pubchem21839 |
PubChem CID |
188444 |
IUPAC Name |
6-bromo-3H-1,3-benzothiazol-2-one |
SMILES |
C1=CC2=C(C=C1Br)SC(=O)N2 |
View More Specs
Products
1
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
AC339880010
SDS
View Documents
|
Thermo Fisher Scientific
339880010
|
1g |
Glass bottle |
N/A
|
N/A
|
|
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
|
|
Description
Specifications
Chemical Identifiers
|
62266-82-4 |
|
230.08 |
|
HECJMTPEVWQFCY-UHFFFAOYSA-N |
|
188444 |
|
C1=CC2=C(C=C1Br)SC(=O)N2 |
|
C7H4BrNOS |
|
MFCD00239363 |
|
2 3h-benzothiazolone, 6-bromo, 2-benzothiazolol, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 6-bromo-2-benzothiazolinone, 6-bromo-2-hydroxy benzothiazol, 6-bromo-3h-benzothiazol-2-one, 6-bromobenzo d thiazol-2 3h-one, pubchem21839 |
|
6-bromo-3H-1,3-benzothiazol-2-one |
Specifications
|
Authentic |
|
White |
|
1g |
|
97.5 |
|
98% |
|
MFCD00239363 |
|
Solubility in water: insoluble in water.,soluble in ethanol |
|
C1=CC2=C(C=C1Br)SC(=O)N2 |
|
230.08 |
|
230.08 |
|
98% |
|
Glass bottle |
|
228°C to 232°C |
|
62266-82-4 |
|
100.0 |
|
C7H4BrNOS |
|
2 3h-benzothiazolone, 6-bromo, 2-benzothiazolol, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 6-bromo-2-benzothiazolinone, 6-bromo-2-hydroxy benzothiazol, 6-bromo-3h-benzothiazol-2-one, 6-bromobenzo d thiazol-2 3h-one, pubchem21839 |
|
HECJMTPEVWQFCY-UHFFFAOYSA-N |
|
6-bromo-3H-1,3-benzothiazol-2-one |
|
188444 |
|
Crystalline Powder |
|
6-Bromo-2-benzothiazolinone, 98% |
SDS