missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-2-benzothiazolinone, 98%

$63.54 - $255.99

Chemical Identifiers

CAS 62266-82-4
Molecular Formula C7H4BrNOS
Molecular Weight (g/mol) 230.079
MDL Number MFCD00239363
InChI Key HECJMTPEVWQFCY-UHFFFAOYSA-N
Synonym 6-bromo-2-benzothiazolinone, 6-bromobenzo d thiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-3h-benzothiazol-2-one, 2 3h-benzothiazolone, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2-hydroxy benzothiazol, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 2-benzothiazolol, 6-bromo, pubchem21839
PubChem CID 188444
IUPAC Name 6-bromo-3H-1,3-benzothiazol-2-one
SMILES C1=CC2=C(C=C1Br)SC(=O)N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6038503 View Documents Alfa Aesar™
H6038503
1g Each for $63.54
 
AAH6038506 View Documents Alfa Aesar™
H6038506
5g Each for $255.99
 
Specifications

Chemical Identifiers

62266-82-4
230.079
HECJMTPEVWQFCY-UHFFFAOYSA-N
188444
C1=CC2=C(C=C1Br)SC(=O)N2
C7H4BrNOS
MFCD00239363
6-bromo-2-benzothiazolinone, 6-bromobenzo d thiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-3h-benzothiazol-2-one, 2 3h-benzothiazolone, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2-hydroxy benzothiazol, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 2-benzothiazolol, 6-bromo, pubchem21839
6-bromo-3H-1,3-benzothiazol-2-one

Specifications

229°C to 233°C
62266-82-4
MFCD00239363
HECJMTPEVWQFCY-UHFFFAOYSA-N
6-bromo-3H-1,3-benzothiazol-2-one
188444
98%
1g
C7H4BrNOS
6-bromo-2-benzothiazolinone, 6-bromobenzo d thiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-3h-benzothiazol-2-one, 2 3h-benzothiazolone, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2-hydroxy benzothiazol, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 2-benzothiazolol, 6-bromo, pubchem21839
C1=CC2=C(C=C1Br)SC(=O)N2
230.079
230.08
6-Bromo-2-benzothiazolinone