missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-2,4-dichloroquinazoline, 97%

Manufacturer:  Alfa Aesar™ H6360306

 View more versions of this product

Catalog No. AAH6360306



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

6-Bromo-2,4-dichloroquinazoline
5g
C8H3BrCl2N2
quinazoline, 6-bromo-2,4-dichloro, 2,4-dichloro-6-bromoquinazoline, 6-bromo-2,4-dichloro-quinazoline, quinazoline,6-bromo-2,4-dichloro, pubchem14690, acmc-1c6nj, abbypharma ap-16-10560, 6-bromanyl-2,4-bis chloranyl quinazoline
C1=CC2=C(C=C1Br)C(=NC(=N2)Cl)Cl
277.93
277.93
97%
White
102393-82-8
MFCD09744007
LBAYOWRVZAKPLS-UHFFFAOYSA-N
6-bromo-2,4-dichloroquinazoline
10107568
Solid

Chemical Identifiers

102393-82-8
277.93
LBAYOWRVZAKPLS-UHFFFAOYSA-N
10107568
C1=CC2=C(C=C1Br)C(=NC(=N2)Cl)Cl
C8H3BrCl2N2
MFCD09744007
quinazoline, 6-bromo-2,4-dichloro, 2,4-dichloro-6-bromoquinazoline, 6-bromo-2,4-dichloro-quinazoline, quinazoline,6-bromo-2,4-dichloro, pubchem14690, acmc-1c6nj, abbypharma ap-16-10560, 6-bromanyl-2,4-bis chloranyl quinazoline
6-bromo-2,4-dichloroquinazoline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents