missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-2(1H)-quinolinone, 96%

Manufacturer:  Alfa Aesar™ H5409514

 View more versions of this product

Catalog No. AAH5409514



Specifications

Specifications

6-Bromo-2(1H)-quinolinone
25g
C9H6BrNO
2 1h-quinolinone, 6-bromo
C1=CC2=C(C=CC(=O)N2)C=C1Br
224.057
224.05
>200°C
1810-66-8
MFCD08703169
YLAFBGATSQRSTB-UHFFFAOYSA-N
6-bromo-1H-quinolin-2-one
12378943
96%

Chemical Identifiers

1810-66-8
224.057
YLAFBGATSQRSTB-UHFFFAOYSA-N
12378943
C1=CC2=C(C=CC(=O)N2)C=C1Br
C9H6BrNO
MFCD08703169
2 1h-quinolinone, 6-bromo
6-bromo-1H-quinolin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents