Please login to your online account to display your discounted pricing

6-Bromo-1H-indazole, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 6-Bromo-1H-indazole
Assay Percent Range 97%
Molecular Formula C7H5BrN2
Formula Weight 197.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC444590010

  SDS
thermo fisher scientific
AC444590010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444590050

  SDS
thermo fisher scientific
AC444590050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 197.04
Quantity 1g
Chemical Name or Material 6-Bromo-1H-indazole
Assay Percent Range 97%
Molecular Formula C7H5BrN2
CAS 79762-54-2