missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromo-1H-indazole, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444590050

 View more versions of this product

Catalog No. AC444590050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

6-Bromo-1H-indazole
5g
96% min. (HPLC)
1h-indazole, 6-bromo, 1h-indazole,6-bromo, 6-bromanyl-1h-indazole, 6-bromo-1-h-indazole, 6-bromo-2h-indazole, 6-bromoindazole, acmc-209pi5, pubchem7831, zlchem 1237
C1=CC2=C(C=C1Br)NN=C2
197.04
197.04
181°C to 185°C
79762-54-2
C7H5BrN2
WMKDUJVLNZANRN-UHFFFAOYSA-N
6-bromo-1H-indazole
17842471
97%

Chemical Identifiers

79762-54-2
197.04
1h-indazole, 6-bromo, 1h-indazole,6-bromo, 6-bromanyl-1h-indazole, 6-bromo-1-h-indazole, 6-bromo-2h-indazole, 6-bromoindazole, acmc-209pi5, pubchem7831, zlchem 1237
6-bromo-1H-indazole
C7H5BrN2
WMKDUJVLNZANRN-UHFFFAOYSA-N
17842471
C1=CC2=C(C=C1Br)NN=C2
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents