missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromo-1H-1,2,3-triazolo[4,5-b]pyridine, TCI America™

$190.97 - $609.59

Chemical Identifiers

CAS 92276-38-5
Molecular Formula C5H3BrN4
Molecular Weight (g/mol) 199.011
InChI Key YKWWQNVTTSHHGH-UHFFFAOYSA-N
Synonym 6-Bromo-4-azabenzotriazole
PubChem CID 494193
IUPAC Name 6-bromo-2H-triazolo[4,5-b]pyridine
SMILES C1=C(C=NC2=NNN=C21)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30341G View Documents TCI America
B30341G
1g Each for $190.97
 
B30345G View Documents TCI America
B30345G
5g Each for $609.59
 
Specifications

Chemical Identifiers

92276-38-5
199.011
6-Bromo-4-azabenzotriazole
6-bromo-2H-triazolo[4,5-b]pyridine
C5H3BrN4
YKWWQNVTTSHHGH-UHFFFAOYSA-N
494193
C1=C(C=NC2=NNN=C21)Br

Specifications

Yellow
1g
C5H3BrN4
YKWWQNVTTSHHGH-UHFFFAOYSA-N
6-bromo-2H-triazolo[4,5-b]pyridine
494193
Crystal-Powder at 20°C
6-Bromo-1H-1,2,3-triazolo[4,5-b]pyridine
206°C
92276-38-5
6-Bromo-4-azabenzotriazole
C1=C(C=NC2=NNN=C21)Br
199.011
199.01
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No