missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromo-1-methyl-1H-indazole, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444600050

 View more versions of this product

Catalog No. AC444600050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

6-Bromo-1-methyl-1H-indazole
5g
96% min. (GC)
1-methyl-6-bromoindazole, 1h-indazole, 6-bromo-1-methyl, 6-bromo-1-methyl-1h-indazole, 6-bromo-1-methyl-indazole, acmc-209m9p, indazole, 6-bromo-1-methyl, pubchem19529
CN1C2=C(C=CC(=C2)Br)C=N1
211.06
211.06
48°C to 50°C
590417-94-0
C8H7BrN2
SVVSOIGNROPKMS-UHFFFAOYSA-N
6-bromo-1-methylindazole
22558675
97%

Chemical Identifiers

590417-94-0
211.06
1-methyl-6-bromoindazole, 1h-indazole, 6-bromo-1-methyl, 6-bromo-1-methyl-1h-indazole, 6-bromo-1-methyl-indazole, acmc-209m9p, indazole, 6-bromo-1-methyl, pubchem19529
6-bromo-1-methylindazole
C8H7BrN2
SVVSOIGNROPKMS-UHFFFAOYSA-N
22558675
CN1C2=C(C=CC(=C2)Br)C=N1
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents