missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromo-1-methyl-1H-indazole, 97%, ACROS Organics™

Chemical Identifiers

CAS 590417-94-0
Molecular Formula C8H7BrN2
Molecular Weight (g/mol) 211.06
InChI Key SVVSOIGNROPKMS-UHFFFAOYSA-N
Synonym 1-methyl-6-bromoindazole, 1h-indazole, 6-bromo-1-methyl, 6-bromo-1-methyl-1h-indazole, 6-bromo-1-methyl-indazole, acmc-209m9p, indazole, 6-bromo-1-methyl, pubchem19529
PubChem CID 22558675
IUPAC Name 6-bromo-1-methylindazole
SMILES CN1C2=C(C=CC(=C2)Br)C=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444600010
SDS View Documents
Thermo Fisher Scientific
AC444600010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC444600050
SDS View Documents
Thermo Fisher Scientific
AC444600050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

590417-94-0
211.06
1-methyl-6-bromoindazole, 1h-indazole, 6-bromo-1-methyl, 6-bromo-1-methyl-1h-indazole, 6-bromo-1-methyl-indazole, acmc-209m9p, indazole, 6-bromo-1-methyl, pubchem19529
6-bromo-1-methylindazole
C8H7BrN2
SVVSOIGNROPKMS-UHFFFAOYSA-N
22558675
CN1C2=C(C=CC(=C2)Br)C=N1

Specifications

Glass Bottle
1g
96% min. (GC)
1-methyl-6-bromoindazole, 1h-indazole, 6-bromo-1-methyl, 6-bromo-1-methyl-1h-indazole, 6-bromo-1-methyl-indazole, acmc-209m9p, indazole, 6-bromo-1-methyl, pubchem19529
CN1C2=C(C=CC(=C2)Br)C=N1
211.06
211.06
6-Bromo-1-methyl-1H-indazole
48°C to 50°C
590417-94-0
C8H7BrN2
SVVSOIGNROPKMS-UHFFFAOYSA-N
6-bromo-1-methylindazole
22558675
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning