Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-1,8-naphthyridin-2(1H)-one, 98%

$108.64 - $373.68

Chemical Identifiers

Chemical Name or Material 6-Bromo-1,8-naphthyridin-2(1H)-one
CAS 72754-05-3
Molecular Formula C8H5BrN2O
Formula Weight 225.05
MDL Number MFCD07781151
Synonym 6-Bromo-2-hydroxy-1,8-naphthyridine; 6-Bromo-1,8-naphthyridin-2-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5004203

  SDS
alfa aesar™
H5004203
1g Each for $373.68

AAH50042MD

  SDS
alfa aesar™
H50042MD
250mg Each for $108.64
Description & Specifications

Specifications

Formula Weight 225.05
Chemical Name or Material 6-Bromo-1,8-naphthyridin-2(1H)-one
Assay 98%
Molecular Formula C8H5BrN2O
MDL Number MFCD07781151
CAS 72754-05-3
Melting Point >300°C
Synonym 6-Bromo-2-hydroxy-1,8-naphthyridine; 6-Bromo-1,8-naphthyridin-2-ol
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
6-Bromo-1,8-naphthyridin-2(1H)-one is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.