missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Bromo-1,4-benzodioxane, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2562514
Description
6-Bromo-1,4-benzodioxane is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Bromo-1,4-benzodioxane is used as a pharmaceutical intermediate.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
6-Bromo-1,4-benzodioxane | |
1.598 | |
>110°C (230°F) | |
1.589 | |
25 g | |
6-bromo-1,4-benzodioxane, 6-bromo-2,3-dihydrobenzo b 1,4 dioxine, 3,4-ethylenedioxybromobenzene, 6-bromo-2,3-dihydro-1,4-benzodioxin, 1,4-benzodioxin, 6-bromo-2,3-dihydro, 4-bromo-1,2-ethylenedioxybenzene, 3,4-ethylendioxybromobenzene, 6-bromo-2,3-dihydro-benzo 1,4 dioxine, pubchem3736, acmc-209kyk | |
C1COC2=C(O1)C=CC(=C2)Br | |
215.046 | |
215.05 |
52287-51-1 | |
259°C to 260°C | |
C8H7BrO2 | |
MFCD00040750 | |
6223 | |
LFCURAJBHDNUNG-UHFFFAOYSA-N | |
6-bromo-2,3-dihydro-1,4-benzodioxine | |
104141 | |
98% |
Chemical Identifiers
52287-51-1 | |
215.046 | |
LFCURAJBHDNUNG-UHFFFAOYSA-N | |
104141 | |
C1COC2=C(O1)C=CC(=C2)Br |
C8H7BrO2 | |
MFCD00040750 | |
6-bromo-1,4-benzodioxane, 6-bromo-2,3-dihydrobenzo b 1,4 dioxine, 3,4-ethylenedioxybromobenzene, 6-bromo-2,3-dihydro-1,4-benzodioxin, 1,4-benzodioxin, 6-bromo-2,3-dihydro, 4-bromo-1,2-ethylenedioxybenzene, 3,4-ethylendioxybromobenzene, 6-bromo-2,3-dihydro-benzo 1,4 dioxine, pubchem3736, acmc-209kyk | |
6-bromo-2,3-dihydro-1,4-benzodioxine |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 257-817-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only