Please login to your online account to display your discounted pricing

6-Bromo-1,4-benzodioxane, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 6-Bromo-1, 4-benzodioxane
CAS 52287-51-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H7BrO2
Formula Weight 215.05
Synonym 6-Bromo-2, 3-dihydro-1, 4-benzodioxine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC441700010

 
thermo fisher scientific
AC441700010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441700050

 
thermo fisher scientific
AC441700050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 215.05
Assay ≥96.0%
Quantity 1g
Color Colorless to pale yellow
Density 1.598g/mL
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 6-Bromo-1, 4-benzodioxane
Assay Percent Range 97%
CAS 52287-51-1
Molecular Formula C8H7BrO2
Infrared Spectrum Authentic
Synonym 6-Bromo-2, 3-dihydro-1, 4-benzodioxine
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.