missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromo[1,3]oxazolo[4,5-b]pyridin-2(3H)-one, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 432680050

 View more versions of this product

Catalog No. AC432680050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

6-Bromo[1, 3]oxazolo[4, 5-b]pyridin-2(3H)-one
96.0
Authentic
Glass bottle
MFCD08689590
6-bromo-3h-oxazolo 4,5-b pyridin-2-one, 6-bromooxazolo 4,5-b pyridin-2 3h-one, 6-bromo-2h,3h-1,3 oxazolo 4,5-b pyridin-2-one, oxazolo 4,5-b pyridin-2 3h-one, 6-bromo, 6-bromo 1,3 oxazolo 4,5-b pyridin-2 3h-one, 6-bromo-oxazolo 4,5-b pyridin-2 3h-one, 6-bromo-3h-1,3 oxazolo 4,5-b pyridin-2-one, pubchem17712, acmc-1cakb, 6-bromooxooxazolo 4,5-b pyridine
C1=C2C(=NC=C1Br)NC(=O)O2
215.01
215.01
21594-52-5
100.0
96% min. (HPLC)
C6H3BrN2O2
5g
VQPBRWIFFBIRRP-UHFFFAOYSA-N
6-bromo-3H-[1,3]oxazolo[4,5-b]pyridin-2-one
13614458
97%

Chemical Identifiers

21594-52-5
215.01
VQPBRWIFFBIRRP-UHFFFAOYSA-N
13614458
C1=C2C(=NC=C1Br)NC(=O)O2
C6H3BrN2O2
MFCD08689590
6-bromo-3h-oxazolo 4,5-b pyridin-2-one, 6-bromooxazolo 4,5-b pyridin-2 3h-one, 6-bromo-2h,3h-1,3 oxazolo 4,5-b pyridin-2-one, oxazolo 4,5-b pyridin-2 3h-one, 6-bromo, 6-bromo 1,3 oxazolo 4,5-b pyridin-2 3h-one, 6-bromo-oxazolo 4,5-b pyridin-2 3h-one, 6-bromo-3h-1,3 oxazolo 4,5-b pyridin-2-one, pubchem17712, acmc-1cakb, 6-bromooxooxazolo 4,5-b pyridine
6-bromo-3H-[1,3]oxazolo[4,5-b]pyridin-2-one
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only