missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-1,2,3,4-tetrahydroisoquinoline, 95%

Manufacturer:  Alfa Aesar™ H6352703

 View more versions of this product

Catalog No. AAH6352703



Specifications

Specifications

6-Bromo-1,2,3,4-tetrahydroisoquinoline
226942-29-6
MFCD07374370
URDGCPQHZSDBRG-UHFFFAOYSA-N
6-bromo-1,2,3,4-tetrahydroisoquinoline
15885183
Solid
1g
C9H10BrN
isoquinoline, 6-bromo-1,2,3,4-tetrahydro, 6-bromo-1,2,3,4-tetrahydro-isoquinoline, acmc-20ailv, 6-bromo-1,2,3,4-tetra hydro iso quinoline, 6-bromo-1,2,3,4-tetrahydroisoquin, 1,2,3,4-tetrahydro-6-bromoisoquinoline, isoquinoline,6-bromo-1,2,3,4-tetrahydro
C1CNCC2=C1C=C(C=C2)Br
212.09
212.09
95%

Chemical Identifiers

226942-29-6
212.09
URDGCPQHZSDBRG-UHFFFAOYSA-N
15885183
C1CNCC2=C1C=C(C=C2)Br
C9H10BrN
MFCD07374370
isoquinoline, 6-bromo-1,2,3,4-tetrahydro, 6-bromo-1,2,3,4-tetrahydro-isoquinoline, acmc-20ailv, 6-bromo-1,2,3,4-tetra hydro iso quinoline, 6-bromo-1,2,3,4-tetrahydroisoquin, 1,2,3,4-tetrahydro-6-bromoisoquinoline, isoquinoline,6-bromo-1,2,3,4-tetrahydro
6-bromo-1,2,3,4-tetrahydroisoquinoline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C

SDS Documents