Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, 97%

$89.14 - $660.35

Chemical Identifiers

Chemical Name or Material 6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene
CAS 27452-17-1
Molecular Formula C14H19Br
Formula Weight 267.21
MDL Number MFCD05664407
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3043603

  SDS
alfa aesar™
H3043603
1g Each for $89.14

AAH3043609

  SDS
alfa aesar™
H3043609
10g Each for $660.35
Description & Specifications

Specifications

Formula Weight 267.21
Chemical Name or Material 6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene
Assay 97%
Molecular Formula C14H19Br
MDL Number MFCD05664407
CAS 27452-17-1
Melting Point 34° to 37°C
Quantity 1g

Applications
6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene is used in Synthesis and evaluation of synthetic retinoid derivatives as inducers of stem cell differentiation.

Solubility
Insoluble in water

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.