Learn More
Sartorius 6 Branch Microsart™ Manifolds
Catalog No. 14558827
Change view
Click to view available options
Type:
Biosart 100 Manifold
Biosart 250 Manifold
Microsart Funnel Manifold
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14558827 Biosart 100 Manifold
14558830 Biosart 250 Manifold
14558823 Microsart Funnel Manifold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14558827 Supplier Sartorius Supplier No. 168M6BS100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Filtration stand designed to adapt to a variety of different needs.
  • 100% stainless steel for minimized cleaning effort
  • Full autoclavability without disassembly
  • Sterile venting in critical applications
  • Reduced risk of secondary contamination
  • Quick connection adapters for fast connection of tubing and fast interconnection of 2 manifolds
  • Low working height for ergonomic working in laminar flow
  • Accurately detects and quantifies microorganisms in liquid samples
  • Method of choice for liquid testing
  • Provides reliable and reproducible results
Specifications
Disposable Reusable
Filter Type Manifold
For Use With (Equipment) Biosart 100 Monitor
Material Stainless Steel
Sterility Non-sterile
Quantity 1
Type Biosart 100 Manifold
Volume (Metric) 100 mL
Length (Metric) 924 mm
Width (Metric) 98 mm
Height (Metric) 120 mm
Show More Show Less